Learn More
Grainger Micro Drill 5/16 Carbide
Catalog No. 502767484
Encompass_Preferred
The Grainger micro drill bits are made of high speed steel and are designed for drilling small, precise holes in metal, plastic, and other materials. They have straight shank ends and are available in a range of sizes from 0.2mm to 3.2mm in diameter. The bits feature 135 degree split points to reduce thrust and wander. They can be used in low RPM, high precision drilling applications. The micro drill bits are sold individually.
Catalog No. 50-276-7484 Supplier Grainger Supplier No. 16T558
The Grainger micro drill bits are made of high speed steel and are designed for drilling small, precise holes in metal, plastic, and other materials. They have straight shank ends and are available in a range of sizes from 0.2mm to 3.2mm in diameter. The bits feature 135 degree split points to reduce thrust and wander. They can be used in low RPM, high precision drilling applications. The micro drill bits are sold individually.