Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Essential Lab Hydrion™ Lo Ion Dispenser Water Test Kit, 5 to 9 pH

Catalog No. 14853159 Shop All Micro Essential Lab Products
Change view
Click to view available options
pH Range:
5 to 9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
14-853-159 5 to 9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-853-159 Supplier Micro Essential Lab Supplier No. LI5900
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

4.57m (15 ft.) Roll of test paper, color chart, 4mL sample tube (capped), protective case (polypropylene)

Gives accurate readings in dilute or unbuffered solutions

For measuring pH of wastewater, pure water, tapwater, rain water, pond water and other dilute solutions where dip-and-read tests may read low.

  • Water pH test range: 5.0 to 9.0
  • Color chart with distinct color matches every 0.5 pH points: 5.0, 5.5, 6.0, 6.5, 7.0, 7.5, 8.0, 8.5, 9.0 pH

Specifications

pH Range 5 to 9
For Use With (Application) For measuring the pH of wastewater, pure water, tap water, rain water and pond water
Includes 15 ft. roll of test paper, matching color chart and one capped 4mL sample tube, packed in a polypropylene protective case
Quantity Each
Type Hydrion™ Lo Ion Dispenser
Length (English) 15 ft.
Length (Metric) 4.57 m
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.