Learn More
Micro Essential Lab Mikro Hydrion™ pH Test Paper
Catalog No. 14853259 Shop All Micro Essential Lab Products
Change view
Click to view available options
pH Range:
0 to 13
0 to 14
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
14-853-259 0 to 13
14-853-160 0 to 14
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-853-259 Supplier Micro Essential Lab Supplier No. M1313
Specifications
pH Range 0 to 13
Includes Double roll, plastic dispenser with stainless steel cutoff lips and two matching color charts, 10 dispenser/carton, 15 ft. roll or test paper
Quantity Each
Type Hydrion™ Micro Dispenser