Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Essential Lab Hydrion™ Sanitizer: Quaternary Test Paper

Catalog No. 15420130 Shop All Micro Essential Lab Products
Change view
Click to view available options
Quantity:
150 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-420-130 150 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 15-420-130 Supplier Micro Essential Lab Supplier No. QC1001
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

  • Hydrion Lo Ion Quat Test Kit: one 15-foot roll of test paper, matching color chart, two sample tubes with caps, an extra roll of test paper and a laminated color chart
  • Hydrion Quat Test Paper: each carton contains 10 kits, each consisting of a 15-foot roll of test paper and matching color chart

Provides a simple, reliable, and economical means to measure quarternary concentration of ammonium, chlorine and iodine sanitizer solutions

Federal, State and Local health regulations require users of Quaternary Ammonium Sanitizer Solutions to have appropriate test kits available to verify the strength of sanitizer solutions. Sanitizer solutions are essential in the food service industry to ensure that sanitizers are at the proper concentration specified by the individual manufacturer.

High Range Quat Test (FS Cat. No. 15-420-130)

  • Measures high range concentrations
  • Color matches at 0-200-400-600-800-100ppm
  • 15 ft. (4.6m) roll
  • 150 tests per roll

Specifications

For Use With (Application) To measure the concentration of quaternary sanitizers, specifically alkyldimethylbenzyl ammonium compounds, Rocall II and Hyamine 3500
Includes One 15 ft. roll of test paper, matching color chart
Quantity 150 Tests
Type Quaternary Test Paper
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.