Learn More
IDEX Health & Science Upchurch LiteTouch Micro Ferrule 10-32 Coned, for 1/32" OD
Catalog No. NC1793765
Encompass_Preferred
LiteTouch MicroFerrule can be used with hard tubing like stainless steel and PEEK Tubing preventing the collapse of tubing ID. This LiteTouch PEEK ferrule has a stainless steel lock ring. It is designed for 1/32"OD tubing. This ferrule is part of a system that helps prevent the twisting of polymer tubing by gripping tubing at two compression points.
Catalog No. NC1793765 Supplier IDEX Health & Science Upchurch Supplier No. LT132
LiteTouch MicroFerrule can be used with hard tubing like stainless steel and PEEK Tubing preventing the collapse of tubing ID. This LiteTouch PEEK ferrule has a stainless steel lock ring. It is designed for 1/32"OD tubing. This ferrule is part of a system that helps prevent the twisting of polymer tubing by gripping tubing at two compression points.