Learn More
MSC Norton Microfiber Polishing Cloth, 16 x 16 in, Yellow
Catalog No. 502838299
Encompass
This microfiber polishing cloth is designed for effective cleaning and debris collection. Its unique fiber structure traps particles, making it suitable for a variety of cleaning tasks, both with and without chemicals. The cloth is highly durable and reusable, maintaining its cleaning abilities through numerous washes.
  • Collects and traps debris effectively.
  • Can be rinsed or machine washed to release embedded particles.
  • Reusable for over 300 washes without losing performance.
  • Suitable for streak-free cleaning of mirrors and optical lenses.
  • Can be used with or without cleaning chemicals.
Catalog No. 50-283-8299 Supplier MSC Supplier No. 41200932
This microfiber polishing cloth is designed for effective cleaning and debris collection. Its unique fiber structure traps particles, making it suitable for a variety of cleaning tasks, both with and without chemicals. The cloth is highly durable and reusable, maintaining its cleaning abilities through numerous washes.
  • Collects and traps debris effectively.
  • Can be rinsed or machine washed to release embedded particles.
  • Reusable for over 300 washes without losing performance.
  • Suitable for streak-free cleaning of mirrors and optical lenses.
  • Can be used with or without cleaning chemicals.