Learn More
Optimize Technologies OPTI-SOLV™ Direct Connect Micro Filter, 10µm, 5pk
SDP
Supplier:  Optimize Technologies 100403707
Encompass_Preferred
For volume critical filtering needs, Optimize offers the  OPTI-SOLV™ Micro Filter. Based on the design that made the Mini Filter successful, the Micro Filter cuts the internal volume to less than 200nL while retaining the ease of use and functionality of the Mini Filter. The Micro Filter is designed to be used in today’s low  volume, high sensitivity chromatographic applications as a last line of defense against debris. As with all Optimize column protection products, the OPTI-SOLV™ Micro Filter features a zero-dead-volume connection utilizing patented floating stem (auto-adjusting) technology. The efficient design requires no holder and threads directly into any 10-32 port. It is available in 5 packs and recommended for volume-critical applications. For use up to 6,000psi.
Catalog No. 50-200-6157