Learn More
World Precision Instrument Micro Fine Mesh Baskets with Lids-WP-4353MM
SDP
Catalog No. 502090216
Encompass
  • Manufactured with stainless steel 304
  • Electrolyte polished
  • Whole and wires size : 1 x1 x 0.5 mm
Catalog No. 50-209-0216 Supplier World Precision Instrument Supplier No. WP4353MM
  • Manufactured with stainless steel 304
  • Electrolyte polished
  • Whole and wires size : 1 x1 x 0.5 mm