Learn More
Electron Microscopy Sciences MicroPoint™ Micro-Forceps Mf-1 Serrated Jaw
SDP
Supplier:  Electron Microscopy Sciences 620861S
Encompass
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Features straight, extra delicate tips for general precision work, 100mm (4.0") in length, tips 0.5mm wide
Type: Serrated Jaw
Catalog No. 50-996-534