Learn More
Electron Microscopy Sciences MicroPoint™ Micro-Forceps Mf-1, Smooth Jaw
SDP
Catalog No. 50996531
Encompass
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Features straight, extra delicate tips for general precision work, 100mm (4.0") in length, tips 0.5mm wide
Type: Smooth Jaw
Catalog No. 50-996-531 Supplier Electron Microscopy Sciences Supplier No. 620851S
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Features straight, extra delicate tips for general precision work, 100mm (4.0") in length, tips 0.5mm wide
Type: Smooth Jaw