Learn More
Electron Microscopy Sciences MicroPoint™ Micro-Forceps Mf-2 Serrated Jaw
SDP
Catalog No. 50996535
Encompass
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Slightly curved jaws designed for precise handling in areas of limited access, Very delicate, 100mm (4.0") in length; 0.5mm width in tips
Type: Serrated Jaw
Catalog No. 50-996-535 Supplier Electron Microscopy Sciences Supplier No. 620862S
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Slightly curved jaws designed for precise handling in areas of limited access, Very delicate, 100mm (4.0") in length; 0.5mm width in tips
Type: Serrated Jaw