Learn More
Electron Microscopy Sciences MicroPoint™ Micro-Forceps Mf-3 Serrated Jaw
SDP
Catalog No. 50996536
Encompass
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Fully curved jaws designed for optimum visibility in precision microscopic work, Very delicate, 100mm (4.0") in length; 0.5mm width in tips
Type: Serrated Jaw
Catalog No. 50-996-536 Supplier Electron Microscopy Sciences Supplier No. 620863S
Micro Forceps/tweezers are designed specifically for applications in the field of microscopic science. Fully curved jaws designed for optimum visibility in precision microscopic work, Very delicate, 100mm (4.0") in length; 0.5mm width in tips
Type: Serrated Jaw