Learn More
IDEX Health & Science Upchurch MICRO FRITZ 1/EA
Catalog No. NC1920562
Encompass_Preferred
Idex Frits: PEEK, Diameter Inner: 0.16 cm, 0.062 in., Porosity: 2 um
Catalog No. NC1920562 Supplier IDEX Health & Science Upchurch Supplier No. A700
Idex Frits: PEEK, Diameter Inner: 0.16 cm, 0.062 in., Porosity: 2 um