Learn More
Chemglass Life Sciences Gas Dispersion Tube, Micro, Extra Coarse Frit
SDP
Catalog No. CG20704
Encompass_Preferred
Gas Dispersion Tube, Micro, Extra Coarse Frit. Micro size gas dispersion tube having a frit 7mm O.D. x 12mm. Stem is 6mm O.D. x 100mm long.
Catalog No. CG20704 Supplier Chemglass Life Sciences Supplier No. CG20704
Gas Dispersion Tube, Micro, Extra Coarse Frit. Micro size gas dispersion tube having a frit 7mm O.D. x 12mm. Stem is 6mm O.D. x 100mm long.