Learn More
Research Products International Corp Micro-Grinder, Pestle Mixer
SDP
Catalog No. 50550334
Encompass
Convenient laboratory tool produces a high-speed mechanical and vortexing action that completely disperses biological samples. Effective for use with pellets, precipitates, other solids and soft tissue. Utilizes disposable or reusable pestles that allow mixing directly inside a 0.5ml or 1.5ml micro-tube. Prevents sample loss and minimizes risk of spills.
Cordless unit uses two AA batteries (included).
Catalog No. 50-550-334 Supplier Research Products International Corp Supplier No. 299220
Convenient laboratory tool produces a high-speed mechanical and vortexing action that completely disperses biological samples. Effective for use with pellets, precipitates, other solids and soft tissue. Utilizes disposable or reusable pestles that allow mixing directly inside a 0.5ml or 1.5ml micro-tube. Prevents sample loss and minimizes risk of spills.
Cordless unit uses two AA batteries (included).