Learn More
Grainger Micro HDMI(M) to HDMI(F) Adapter 6Inch
Supplier:  Grainger 14C364
Encompass_Preferred
The Monoprice HDMI Adapter is a passive adapter that converts an HDMI signal to a DVI signal. It allows you to connect an HDMI source device such as a Blu-ray player, game console, or laptop to a DVI display or projector. This adapter supports resolutions up to 1920x1200 and 1080p Full HD. It has 24k gold-plated connectors for optimal signal transfer. The adapter is compact and portable. It does not require an external power source. This is a simple plug-and-play solution for connecting HDMI to DVI.
Catalog No. 50-274-8992