Learn More
Electron Microscopy Sciences Micro Hook, Stainless Steel
SDP
Catalog No. 50996709
Encompass
Micro tools are forged from the finest quality stainless steel and are precision hand ground, lapped, and polished. These tools are well suited for most applications in the field of microscopic science which demands the ultimate in precision miniature tools. Micro-Hook, guarded type, anti-snag guard, 0.3mm. ID, 1mm OD including clasp.
redo select columns with descriptions: Stainless Steel
Catalog No. 50-996-709 Supplier Electron Microscopy Sciences Supplier No. 6209606
Micro tools are forged from the finest quality stainless steel and are precision hand ground, lapped, and polished. These tools are well suited for most applications in the field of microscopic science which demands the ultimate in precision miniature tools. Micro-Hook, guarded type, anti-snag guard, 0.3mm. ID, 1mm OD including clasp.
redo select columns with descriptions: Stainless Steel