Learn More
Fine Science Tools Micro Knives - Plastic Handle, 22.5° Cutting Angle, 13.5 cm
SDP
Catalog No. NC9864398
Encompass
Product: Micro Knife - Plastic Handle/22.5 deg. Cutting Angle/13.5cm/ Blade Shape: 22.5 deg. Cutting Angle/ Cutting Edge: 4.5mm/ Thickness: 0.15mm/ Alloy / Material: Carbon Steel Tip / Plastic Handle
Catalog No. NC9864398 Supplier Fine Science Tools Supplier No. 1031614
Product: Micro Knife - Plastic Handle/22.5 deg. Cutting Angle/13.5cm/ Blade Shape: 22.5 deg. Cutting Angle/ Cutting Edge: 4.5mm/ Thickness: 0.15mm/ Alloy / Material: Carbon Steel Tip / Plastic Handle