Learn More
Fine Science Tools Micro Knives - Plastic Handle, Needle Blade, 14.5 cm
SDP
Catalog No. NC9691655
Encompass
Product: Micro Knife - Plastic Handle/Needle Blade/14.5cm/ Blade Shape: Needle Blade/ Cutting Edge: 4mm/ Thickness: 0.2mm/ Alloy / Material: Carbon Steel Tip / Plastic Handle
Catalog No. NC9691655 Supplier Fine Science Tools Supplier No. 1031814
Product: Micro Knife - Plastic Handle/Needle Blade/14.5cm/ Blade Shape: Needle Blade/ Cutting Edge: 4mm/ Thickness: 0.2mm/ Alloy / Material: Carbon Steel Tip / Plastic Handle