Learn More
Roboz Surgical MICRO LIG NDLE 475IN SHP LEFT
SDP
Catalog No. 5000574519
Encompass
This micro ligature needle is designed for precise surgical procedures, specifically for pulling suture material around vessels or nerves. Measuring 4.75 in. in total length, the instrument features a left-curved geometry and a sharp, pointy tip engineered to facilitate tissue penetration.
  • Sharp, pointy tip for efficient tissue penetration
  • Left-curved design requires clockwise insertion and counter-clockwise withdrawal
  • Total length of 4.75 in.
  • Suitable for vascular and nerve manipulation
Catalog No. 50-005-74519 Supplier Roboz Surgical Supplier No. RS6202
This micro ligature needle is designed for precise surgical procedures, specifically for pulling suture material around vessels or nerves. Measuring 4.75 in. in total length, the instrument features a left-curved geometry and a sharp, pointy tip engineered to facilitate tissue penetration.
  • Sharp, pointy tip for efficient tissue penetration
  • Left-curved design requires clockwise insertion and counter-clockwise withdrawal
  • Total length of 4.75 in.
  • Suitable for vascular and nerve manipulation