Learn More
Research Products International Corp Flea Micro Magnetic Stirring Bar, 7 mm x 2 mm
SDP
Catalog No. 50999100
Encompass
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers. Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.
This item is not returnable. View return policy
Catalog No. 50-999-100 Supplier Research Products International Corp Supplier No. F371190007
Micro (Flea) magnetic stirring bars are designed for stirring small volumes in vessels such as vials, tubes and gradient makers. Particularly useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated.