Learn More
Valley Forge Tape And Label Co Inc Micromate Yellow Labels, 500 pk
Catalog No. NC9142470
Encompass
This item has a minimum order quantity of 3 per supplier requirements. Label; Micromate; Endure extreme temp, Autoclaving, PCR Cycling in Oil will not come off in Rotors, Cyclers/Racks; Blue, Green, Red, Yellow, White; Matte surface suitable for Pencil, Pen, Marker; Fits 1.5mL Centrifuge Tubes; 500/ST
Catalog No. NC9142470 Supplier Valley Forge Tape And Label Co Inc Supplier No. 6509
This item has a minimum order quantity of 3 per supplier requirements. Label; Micromate; Endure extreme temp, Autoclaving, PCR Cycling in Oil will not come off in Rotors, Cyclers/Racks; Blue, Green, Red, Yellow, White; Matte surface suitable for Pencil, Pen, Marker; Fits 1.5mL Centrifuge Tubes; 500/ST