Learn More
Scientific Instrument Services Inc MICRO-MESH
Catalog No. NC9744888
Encompass
This item has a minimum qty of 6 per supplier requirements.
Micromesh Abrasives,4 in. x 6 in. sheet, 6000 Grit,color:Orange,can be used either wet or dry,have long life, since the crystals will continue to cut effectively until they eventually become dull and lose their sharp cutting edges
Catalog No. NC9744888 Supplier Scientific Instrument Services Inc Supplier No. MM6000
This item has a minimum qty of 6 per supplier requirements.
Micromesh Abrasives,4 in. x 6 in. sheet, 6000 Grit,color:Orange,can be used either wet or dry,have long life, since the crystals will continue to cut effectively until they eventually become dull and lose their sharp cutting edges