Learn More
MilliporeSigma™ Supelco™ Micro-Metering Valve (Low Pressure)
Catalog No. 111004880 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Size:
1/4-28 Flangeless Fittings
10-32 Fingertight Fittings
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
11-100-4880 1/4-28 Flangeless Fittings
11-100-4777 10-32 Fingertight Fittings
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4880 Supplier MilliporeSigma™ Supelco™ Supplier No. 502367
Add to Cart
Add to Cart
Designed for the control of fluid flow rates
  • Maximum Pressure: 800 psi
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Application) Peristaltic Pumps, Pump Fluid-Transfer, Mass Spectrometry and Fraction Collection
Material PEEK
Type Low Pressure
Size 1/4-28 Flangeless Fittings