Learn More
Glass Expansion MicroMist Nebulizer 0.4mL/min with EzyFit & EzyLok & 0.5 x 1.6 x 700mm Tube
SDP
Catalog No. NC2054084
Encompass_Preferred
MicroMist Nebulizer 0.4 mL/min, includes EzyFit Connector with 0.50 mm ID x 1.6 mm OD x 700 mm long tubing (1 piece) and EzyLok Connector (1 piece).Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 0.4 mL/min. Recommended not to exceed optimum 0.4 mL/min sample flow rate; however, lower gas and sample flow rates are OK. Backpressure should read ~35psi at 1.0 L/min gas flow rate. Tolerates up to 15% TDS. MicroMist is the standard configuration for many ICP-MS manufacturers due to its excellent sensitivity and precision.Replacement parts include P/Ns EZT-16-50 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) MicroMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2054084 Supplier Glass Expansion Supplier No. AR351FM04EX
MicroMist Nebulizer 0.4 mL/min, includes EzyFit Connector with 0.50 mm ID x 1.6 mm OD x 700 mm long tubing (1 piece) and EzyLok Connector (1 piece).Optimum nebulizer gas flow rate is 1.0 L/min and sample flow rate is 0.4 mL/min. Recommended not to exceed optimum 0.4 mL/min sample flow rate; however, lower gas and sample flow rates are OK. Backpressure should read ~35psi at 1.0 L/min gas flow rate. Tolerates up to 15% TDS. MicroMist is the standard configuration for many ICP-MS manufacturers due to its excellent sensitivity and precision.Replacement parts include P/Ns EZT-16-50 and 502702994.Recommend nebulizer maintenance tools, Fluka RBS25 (P/N NC9000117) and Eluo Nebulizer Cleaning Tool (P/N NC9462559). When nebulizer is not in use, best to store in the provided hard plastic case.Consider upgrading to our new Direct Connect (DC) MicroMist to take advantage of the cost-effective design improvements.Glass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001