Learn More
Fine Science Tools Micro-Mosquito Hemostats, Stainless Steel, Serrated, 12 cm
SDP
Catalog No. NC1464663
Encompass
Tip Shape: Angled 90 Degrees/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12cm
This item is not returnable. View return policy
Catalog No. NC1464663 Supplier Fine Science Tools Supplier No. 1301212
Tip Shape: Angled 90 Degrees/ Tips: Serrated/ Tip Width: 1.3mm/ Clamping Length: 20mm/ Alloy / Material: Stainless Steel/ Length: 12cm