Learn More
Sklar Instruments Micro Mosquito Forceps, Sklar, 5 in., Curved, Smooth, Delicate, Stainless Steel. Sklar Halsted Micro Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. EA/1.
SDP
Catalog No. 501437390 Shop All Sklar Instruments Products
Encompass_Preferred
Micro Mosquito Forceps, Sklar, 5 in., Curved, Smooth, Delicate, Stainless Steel. Sklar Halsted Micro Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. They are available curved or straight. The standard length for this instrument is 5 inches, although Sklar does offer some with a length of 5-1/2 inches. Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate. This product is curved with smooth tips and a length of 5 inches.
This item is not returnable. View return policy
Catalog No. 50-143-7390 Supplier Sklar Instruments Supplier No. 171225
Micro Mosquito Forceps, Sklar, 5 in., Curved, Smooth, Delicate, Stainless Steel. Sklar Halsted Micro Mosquito Forceps are used in multiple procedures as a hemostat for small blood vessels. The ratcheted finger ring handle provides a secure, locking grip to halt blood flow. They are not appropriate for use with large vessels or arteries. A toothed variation is available for grasping delicate tissue in skin grafting, biopsies, or ophthalmologic procedures. Mosquito forceps are known for their fine tips, and short, fully serrated jaws. They are available curved or straight. The standard length for this instrument is 5 inches, although Sklar does offer some with a length of 5-1/2 inches. Halsted Mosquito Forceps are very similar to Hartmann Mosquito Forceps, however, the Hartmann style is finer and more delicate. This product is curved with smooth tips and a length of 5 inches.