Learn More
World Precision Instrument Micro Mosquito Hemostatic Forceps, 12.5cm, Right angle, Serrated
SDP
Catalog No. 502539491
Encompass
Micro Mosquito Forceps, also known as microsurgical mosquito forceps or micro hemostatic forceps, are designed for extremely delicate and precise surgical procedures. Micro Mosquito Forceps are significantly smaller in size compared to regular mosquito forceps. These are 12.5 cm (5") long with fine tips. Looking at the end of the tip of one blade of these forceps, the dimensions are 0.4 mm thick and 0.8 mm wide.
Catalog No. 50-253-9491 Supplier World Precision Instrument Supplier No. 503360
Micro Mosquito Forceps, also known as microsurgical mosquito forceps or micro hemostatic forceps, are designed for extremely delicate and precise surgical procedures. Micro Mosquito Forceps are significantly smaller in size compared to regular mosquito forceps. These are 12.5 cm (5") long with fine tips. Looking at the end of the tip of one blade of these forceps, the dimensions are 0.4 mm thick and 0.8 mm wide.