Learn More
Hampton Research Micro-Tools Set
SDP
Supplier:  Hampton Research HR4827
Encompass
Micro-Tools™ are the smallest available precision instruments for laboratory use. These tools are realistically proportioned for macromolecular and small molecule crystallization work. The high quality, precision Micro-Tools ease microscopic manipulations. The eight specifically engineered Micro-Tools are useful in a variety of applications in the crystallization lab. Eight tools, a handle and wood storage box. Please note items from this supplier are non-returnable.
Catalog No. 50-008-7