Learn More
Electron Microscopy Sciences Micro Needle, 45 degree , Stainless Steel
SDP
Catalog No. 50996705
Encompass
Micro tools are forged from the finest quality stainless steel and are precision hand ground, lapped, and polished. These tools are well suited for most applications in the field of microscopic science which demands the ultimate in precision miniature tools. Micro-Needle, bent 45 degree, bent section is 5mm long, sharp tip.
redo select columns with descriptions: Stainless Steel
Catalog No. 50-996-705 Supplier Electron Microscopy Sciences Supplier No. 6209602
Micro tools are forged from the finest quality stainless steel and are precision hand ground, lapped, and polished. These tools are well suited for most applications in the field of microscopic science which demands the ultimate in precision miniature tools. Micro-Needle, bent 45 degree, bent section is 5mm long, sharp tip.
redo select columns with descriptions: Stainless Steel