Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Stoelting™ Micro Needle Holder with Lock
SDP

Catalog No. 10001676 Shop All Stoelting Co Products
Change view
Click to view available options
Length (English):
5.125 in.
7.125 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English)
10-001-676 7.125 in.
10-001-675 5.125 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 10-001-676 Supplier Stoelting™ Supplier No. 5213539
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Stoelting™ Micro Needle Holder with Lock is designed to give precise control for various needle types. Features include: Stainless steel

Specifications

For Use With (Application) Neuroscience
Length (English) 7.125 in.
Width (Metric) 0.6 mm tip
Width (English) 0.6 mm tip
Model Curved with lock
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.