Learn More
Trajan Scientific MICRO NEEDLE VALVE MOVPT-1/50
SDP
Catalog No. 502530154
Encompass_Preferred
Trajan Micro Control Valve and Accessories, Valve Code: MOVPT, Length Stem: 50 mm, 1.9 in., Style: Pneumatic T Dual Outlet, Connection Size: 1/16 in., 0.15 cm, Stainless Steel, Max. Pressure: 700 psi, Micro control valve, Temperature Maximum: 572 deg.F, Temperature Max: 300 deg.C
Catalog No. 50-253-0154 Supplier Trajan Scientific Supplier No. 1236103
Trajan Micro Control Valve and Accessories, Valve Code: MOVPT, Length Stem: 50 mm, 1.9 in., Style: Pneumatic T Dual Outlet, Connection Size: 1/16 in., 0.15 cm, Stainless Steel, Max. Pressure: 700 psi, Micro control valve, Temperature Maximum: 572 deg.F, Temperature Max: 300 deg.C