Learn More
Cole-Parmer AEMC 6240 10-Amp Micro-Ohmmeter, 1µ Ohms to 400 Ohms
Supplier:  Cole-Parmer 2744345
Encompass
This micro-ohmmeter features reliable low resistance measurements. Its four-terminal Kelvin resistance measurement eliminates test lead resistance. It has a 10A test current up to 4000, 0.25 percent basic accuracy and one micro-ohm resolution.
Catalog No. 50-627-325