Learn More
Bellco Glass, Inc. Micro-Orbital Shaker,12V-115V
SDP
Catalog No. 501946165
Encompass_Preferred
The Bellco Benchtop 12 V Micro Orbital Shakers are ideal for both rigid polystyrene and flexible polyvinyl 96-well micro tube plates commonly used in cell culture, virology, immunology and serology. They are engineered to operate between the speed range of 60 to 1150 rpm and have a 5 minute timer for timed mixing continuous operation. The true orbital motion results in increased mixture homogeneity compared with oscillating or vibrating shakers.. MADE IN THE USA
Catalog No. 50-194-6165 Supplier Bellco Glass, Inc. Supplier No. 774510115
The Bellco Benchtop 12 V Micro Orbital Shakers are ideal for both rigid polystyrene and flexible polyvinyl 96-well micro tube plates commonly used in cell culture, virology, immunology and serology. They are engineered to operate between the speed range of 60 to 1150 rpm and have a 5 minute timer for timed mixing continuous operation. The true orbital motion results in increased mixture homogeneity compared with oscillating or vibrating shakers.. MADE IN THE USA