Learn More
Allied High Tech Products Micro Organic Soap, 32 oz
SDP
Catalog No. NC1804291
Encompass
A high-purity cleaning solution for removing micro-contaminants and polishing solutions from samples prior to microscopic examination. Mixed with water at 2-5% by volume.
Catalog No. NC1804291 Supplier Allied High Tech Products Supplier No. 14810000
A high-purity cleaning solution for removing micro-contaminants and polishing solutions from samples prior to microscopic examination. Mixed with water at 2-5% by volume.