Learn More
Bellco Glass, Inc. Micro-Oven,115V w/4 Position Rotisserie
SDP
Catalog No. 501946521
Encompass_Preferred
The Bellco Auto-blot Micro Hybridization Oven consists of an incubator with an integral rotisserie device. The rotisserie has the capacity for up to 4 large hybridization bottles or 8 small hybridization bottles. Alternatively, any combination of large or small bottles can be used. This Auto-blot Micro Hybridization Oven features an advanced temperature controller for precise control of oven temperature. Ext: 14”x17”x13” (35x42x32cm) 115v. MADE IN THE USA
Catalog No. 50-194-6521 Supplier Bellco Glass, Inc. Supplier No. 793000110
The Bellco Auto-blot Micro Hybridization Oven consists of an incubator with an integral rotisserie device. The rotisserie has the capacity for up to 4 large hybridization bottles or 8 small hybridization bottles. Alternatively, any combination of large or small bottles can be used. This Auto-blot Micro Hybridization Oven features an advanced temperature controller for precise control of oven temperature. Ext: 14”x17”x13” (35x42x32cm) 115v. MADE IN THE USA