Learn More
Chemglass Life Sciences Micro Overhead Stirrer for Tissue Grinder
SDP
Catalog No. 501531410
Encompass_Preferred
Micro Overhead Stirrer for Tissue Grinder. This lightweight micro stirrer is ideal for use with tissue grinders having Stainless Steel pestles. Supplied complete with a stirrer/controller, tripod with a vertical support rod, clamp holder and a coupling to fit the 6.35mm diameter of the Stainless Steel pestle rod.
Catalog No. 50-153-1410 Supplier Chemglass Life Sciences Supplier No. CG2039A500
Micro Overhead Stirrer for Tissue Grinder. This lightweight micro stirrer is ideal for use with tissue grinders having Stainless Steel pestles. Supplied complete with a stirrer/controller, tripod with a vertical support rod, clamp holder and a coupling to fit the 6.35mm diameter of the Stainless Steel pestle rod.