Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Particles Based on Polymethacrylate, Analytical Standard, Size: 1 m, MilliporeSigma™ Supelco™

Catalog No. 111004255 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4255 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-4255 Supplier MilliporeSigma Supelco Supplier No. 908755MLF
Add to Cart
Add to Cart

Micro particles based on polymethacrylate may exhibit mucoadhesive properties and hence can show potential applications in developing oral insulin delivery systems.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Particles
Concentration 10% (solids)
MDL Number MFCD00198073
Synonym 2-Propenoic acid,2-methyl homopolymer; 2-methyl-2-propenoic acid homopolymer; Microparticles based on PMAA; Microparticles based on poly(methacrylic acid); Polymethacrylate
Quantity 5 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.