Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Particles Based on Polystyrene, Analytical Standard, Size: 30 m, MilliporeSigma™ Supelco™

Catalog No. 111004082 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4082 5 mL
11-100-3815 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4082 Supplier MilliporeSigma Supelco Supplier No. 841355MLF
Add to Cart
Add to Cart

Specifications

Density 1.05 g/cm3
Physical Form Particles
Concentration 10% (solids)
MDL Number MFCD00243243
Synonym Latex beads from PS
Quantity 5 mL
Grade Analytical Standard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.