Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Particles Based on Polystyrene, Dark blue, Size: 10 m, MilliporeSigma™ Supelco™

Catalog No. 111003849 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-3849 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-3849 Supplier MilliporeSigma Supelco Supplier No. 554635MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Concentration 5% (solids)
Content And Storage 2°C to 8°C
Quantity 5 mL
Synonym Latex beads from PS, dark blue
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.