Learn More
Micro particles based on polystyrene, dark blue size: 3 m, MilliporeSigma™ Supelco™
Shop All MilliporeSigma Supelco ProductsSpecifications
Specifications
| Physical Form | Aqueous Suspension |
| Concentration | 5% (solids) |
| MDL Number | MFCD00131491 |
| Synonym | Latex beads from PS, dark blue |
| Type | Organic Standard |
| Quantity | 5 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.