Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Micro Particles Based on Polystyrene, Size: 5 m, MilliporeSigma™ Supelco™

Catalog No. 111004010 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-100-4010 5 mL
11-100-3777 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-100-4010 Supplier MilliporeSigma Supelco Supplier No. 796335MLF
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Concentration 10% (solids)
Content And Storage 2°C to 8°C
Quantity 5 mL
Synonym Latex beads from PS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.