Learn More
Thomas Scientific Micro Pestle, PP, 12/pk
SDP
Catalog No. NC1795366
Encompass
This double-ended polypropylene micro pestle features one end that fits 1.5ml microcentrifuge tubes and another end that fits 0.5ml tubes. Autoclavable. Micro pestles are excellent for resuspending bacterial cells and precipitates of nucleic acids or proteins, and are useful for tissue grinding.
Catalog No. NC1795366 Supplier Thomas Scientific Supplier No. 1194N23
This double-ended polypropylene micro pestle features one end that fits 1.5ml microcentrifuge tubes and another end that fits 0.5ml tubes. Autoclavable. Micro pestles are excellent for resuspending bacterial cells and precipitates of nucleic acids or proteins, and are useful for tissue grinding.