Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO™ Micro pH Electrodes

Catalog No. 01911475 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Length (Metric):
40 mm
60 mm
100 mm
130 mm
180 mm
230 mm
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (Metric)
01-911-475 100 mm
01-911-477 60 mm
01-911-476 230 mm
01-917-147 130 mm
01-911-442 180 mm
01-914-306 40 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 01-911-475 Supplier METTLER TOLEDO™ Supplier No. 51343165
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

pH electrodes designed for small sample volumes or for measuring in small containers.

  • For use with METTLER TOLEDO SevenExcellence and SevenCompact meters as well as other brands. (Connection cable is not included; sold separately.)
  • Intelligent Sensor Management (ISM) electrodes capture and save calibration data, which is then automatically recognized by the meter once connected.
  • Capture electrode calibration and results with LabX software, designed for advanced data management and 21CFR part 11 regulatory compliance.
  • Compatible with InMotion Autosampler to automate measurement and cleaning of up to 303 samples.

Featured Applications

  • Vials
  • Microplates
  • Test tubes
  • Serum and Gastric juice
  • Micro-biological samples
  • Paint
  • Gel
  • Soap
  • Shampoo
  • Cosmetics

Electrode Specifications

  • InLab Micro: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Micro Pro-ISM: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Ultra-Micro-ISM: ceramic junction; refillable with FRISCOLYT-B™ (51343185) - keep probe filled with electrolyte for accurate results
  • InLab Semi-Micro: ceramic junction; non-refillable - replace when electrolyte depleted
  • InLab Semi-Micro-L: ceramic junction; refillable with 3M KCl (51350072) - keep probe filled with electrolyte for accurate results
  • InLab Flex-Micro: porous PTFE junction; non-refillable - replace when electrolyte depleted

Accessories & Care

  • Store in 3M KCl (51350072) or InLab Storage Solution (30111142)
  • Recommended for use with METTLER TOLEDO buffers (Sachets: 51302080, Bottles: 63056116)
TRUSTED_SUSTAINABILITY

Specifications

Connector Types S7
Junction Open
Material Glass
pH Range 0 to 12
Temperature (Metric) Operating 0°C to 100°C
Type Combination Electrode
Filling Solution Type Xerolyt Extra Polymer
For Use With (Application) Measure pH in the small or narrow neck vessels
For Use With (Equipment) Requires Cable, S7 to BNC (30281915)
Internal Reference ARGENTHAL
Membrane A41 Glass
Model InLab™ Semi-Micro
Response Time <20 sec.
Storage Requirements FRISCOLYT-B
Temperature (English) Operating 32°F to 212°F
Temperature (Metric) Range 0°C to 100°C
Length (English) 3.93 in.
Length (Metric) 100 mm
Diameter (English) 0.23 in.
Diameter (Metric) 6 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.