Learn More
Research Products International Corp Micro-Plate Incubator Shaker, Two Plate Capacity
SDP
Supplier:  Research Products International Corp 270400
Encompass
These incubators provide uniform and reliable mixing and heating for two or four micro-plates. Digital control pad allows independent setting of temperature, shaking speed and duration. Unique double chamber design and heated lid provide uniform heating and temperature stability. Accommodates all standard 96 and 384 well plates.
Catalog No. 50-197-6846