Learn More
Verichem Laboratories MICRO PROTEIN STD KIT
Catalog No. NC0006353
Encompass_Preferred
Intended for calibration or calibration verification. Liquid, ready-to-use standards. Human protein components for optimum reactivity. Extremely stable formulation with 24 month shelf life at 2 to 8 degrees C. Certified gravimetric concentrations with exceptional lot-to-lot duplication Suitable for any turbidimetric or colormetric method Azide free
Catalog No. NC0006353 Supplier Verichem Laboratories Supplier No. 9480
Intended for calibration or calibration verification. Liquid, ready-to-use standards. Human protein components for optimum reactivity. Extremely stable formulation with 24 month shelf life at 2 to 8 degrees C. Certified gravimetric concentrations with exceptional lot-to-lot duplication Suitable for any turbidimetric or colormetric method Azide free