Learn More
Starna Cells Inc MICRO QUARTZ CELL 10MM THK BAS
SDP
Catalog No. NC9669262
Encompass_Preferred
Micro, w/lid, Self-masking, Black walls, Spectrosil quartz, 2W Internal, 12.5L x 12.5W x 45H, 0.580mL, Open top, supplied with non-sealing PTFE cover, Two polished windows, Walls polished internally, Fine ground externally
Catalog No. NC9669262 Supplier Starna Cells Inc Supplier No. 18B9Q10
Micro, w/lid, Self-masking, Black walls, Spectrosil quartz, 2W Internal, 12.5L x 12.5W x 45H, 0.580mL, Open top, supplied with non-sealing PTFE cover, Two polished windows, Walls polished internally, Fine ground externally