Learn More
Abraxis ABRAXIS Microcystins Strip Test, 20 tests, Immunochromatographic, Recreational Water, Detection Range 0-10 ppb
SDP
Catalog No. NC1394971
Encompass
ABRAXIS Microcystins Strip Test, 20 tests, Immunochromatographic, Recreational Water, Detection Range 0-10 ppb
Catalog No. NC1394971 Supplier Abraxis Supplier No. 520022
ABRAXIS Microcystins Strip Test, 20 tests, Immunochromatographic, Recreational Water, Detection Range 0-10 ppb