Learn More
Braintree Scientific MICRO-RENATH IMP TUB .010X.005
SDP
Catalog No. NC9026118
Encompass
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**
Catalog No. NC9026118 Supplier Braintree Scientific Supplier No. MRE01050
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**