Learn More
Braintree Scientific MICRO-RENATHANE .033X.014 100F
SDP
Catalog No. NC0546443
Encompass
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**
Catalog No. NC0546443 Supplier Braintree Scientific Supplier No. MRE033100FT
In our opinion, Micro-Renathane is the most blood compatible tubing ever made. For implantation studies in experimental animals this tubing offers extended catheter life and reduces the probability of intravascular thrombosis. **Sold for research purposes only and not for human use**